Skip to Content
Merck
vibrant-m

Zileuton Related Compound A

vibrant-m
Synonym(s):
N-(1-Benzo[b]thien-2-ylethyl)urea, N-Dehydroxyzileuton
Empirical Formula (Hill Notation):
C11H12N2OS
CAS Number:
Molecular Weight:
220.29

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1