Trizma® maleate
Synonym(s):
TRIS maleate salt, Tris(hydroxymethyl)aminomethane maleate salt, Tromethamine maleate salt
Linear Formula:
NH2C(CH2OH)3 · C4H4O4
Products
Shipping
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing