Tributyl acetylcitrate
Synonym(s):
Tributyl 2-acetylcitrate, Acetyl tributyl citrate, Tributylacetyl citrate
Linear Formula:
CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
402.48
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing