Skip to Content
Merck
trans-2-Phenylcyclopropylamine hydrochloride 97%

trans-2-Phenylcyclopropylamine hydrochloride

trans-2-Phenylcyclopropylamine hydrochloride 97%
Synonym(s):
Tranylcypromine
Linear Formula:
C6H5C3H4NH2·HCl
CAS Number:
Molecular Weight:
169.65

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing