![Thiamet G TMG](/deepweb/assets/sigmaaldrich/product/structures/175/263/50532256-02c7-4a95-9b06-794eeadfa9b6/640/50532256-02c7-4a95-9b06-794eeadfa9b6.png)
Thiamet G
![Thiamet G TMG](/deepweb/assets/sigmaaldrich/product/structures/175/263/50532256-02c7-4a95-9b06-794eeadfa9b6/640/50532256-02c7-4a95-9b06-794eeadfa9b6.png)
Synonym(s):
(3aR,5R,6S,7R,7aR)-2-(ethylamino)-3a,6,7,7a-tetrahydro-5-(hydroxymethyl)-5H-Pyrano[3,2-d]thiazole-6,7-diol
Empirical Formula (Hill Notation):
C9H16N2O4S
CAS Number:
Molecular Weight:
248.30
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing