
τ-Fluvalinate

Synonym(s):
N-[2-Chloro-4-(trifluoromethyl)phenyl]-D-valine (RS)-cyano(3-phenoxyphenyl)methyl ester
Empirical Formula (Hill Notation):
C26H22ClF3N2O3
CAS Number:
Molecular Weight:
502.91
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2