
Tetraphenylphosphonium iodide

Synonym(s):
Tetraphenylphosphonium iodide
Empirical Formula (Hill Notation):
C24H20IP
CAS Number:
Molecular Weight:
466.29
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing