Skip to Content
Merck
tert-Butylmagnesium chloride solution 2.0 M in diethyl ether

tert-Butylmagnesium chloride solution

tert-Butylmagnesium chloride solution 2.0 M in diethyl ether
Linear Formula:
(CH3)3CMgCl
CAS Number:
Molecular Weight:
116.87

Products

Shipping
Product Category
Formula Weight
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2