Skip to Content
Merck
Terazosin Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Terazosin hydrochloride

Terazosin Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Terazosin hydrochloride dihydrate, 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonyl]piperazine hydrochloride dihydrate
Empirical Formula (Hill Notation):
C19H25N5O4 · HCl · 2H2O
CAS Number:
Molecular Weight:
459.92

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique