Skip to Content
Merck
Teniposide Related Compound A certified reference material, pharmaceutical secondary standard

Teniposide Related Compound A

Teniposide Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
4′-Demethylepipodophyllotoxin, (5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydrofuro[3′,4′:6,7]naphtho[2,3-d][1,3]dioxol-6(5aH)-one
Empirical Formula (Hill Notation):
C21H20O8
CAS Number:
Molecular Weight:
400.38

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade