Skip to Content
Merck
TCS-1102 ≥98% (HPLC)

TCS-1102

TCS-1102 ≥98% (HPLC)
Synonym(s):
BBAC, (2S)-N-[1,1′-Biphenyl]-2-yl-1-[2-[(1-methyl-1H-benzimidazol-2-yl)thio]acetyl]-2-pyrrolidinecarboxamide
Empirical Formula (Hill Notation):
C27H26N4O2S
CAS Number:
Molecular Weight:
470.59

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing