Skip to Content
Merck
Sodium L-lactate-3-13C solution endotoxin tested, 45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP)

Sodium L-lactate-3-13C solution

Sodium L-lactate-3-13C solution endotoxin tested, 45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP)
Synonym(s):
L-Lactic acid-3-13C sodium salt solution
Linear Formula:
13CH3CH(OH)CO2Na
CAS Number:
Molecular Weight:
113.05

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1