Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium dithionite
Synonym(s):
Sodium dithionite, Sodium hydrosulfite, Sodium hypodisulfite
Linear Formula:
NaO
2
SSO
2
Na
CAS Number:
7775-14-6
Molecular Weight:
174.11
EC Number:
231-890-0
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (1)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.06505
Sodium dithionite,
EMPLURA
®
Expand
View Pricing
1.06507
Sodium dithionite,
for analysis EMSURE
®
Expand
View Pricing
28-2930
Sodium dithionite,
CP
Expand
View Pricing