
Riluzole

Synonym(s):
2-Amino-6-(trifluoromethoxy)benzothiazole, Riluzole
Empirical Formula (Hill Notation):
C8H5F3N2OS
CAS Number:
Molecular Weight:
234.20
Products
Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing