Skip to Content
Merck
Reinforced Membrane Filter Millipore, filter diam. 47 mm, hydrophilic

Reinforced Membrane Filter

Reinforced Membrane Filter Millipore, filter diam. 47 mm, hydrophilic
Synonym(s):
MCE Membrane Filter, 1.2 μm Pore Size

Products

Product Category
Brand
Manufacturer
Diameter (mm)
Quality Segment
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing