Skip to Content
Merck
Pyridate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Pyridate

Pyridate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
O-(6-Chloro-3-phenylpyridazin-4-yl) S-octyl carbonothioate
Empirical Formula (Hill Notation):
C19H23ClN2O2S
CAS Number:
Molecular Weight:
378.92

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique