Skip to Content
Merck
Proguanil Related Compound E pharmaceutical secondary standard, certified reference material

Proguanil Related Compound E

Proguanil Related Compound E pharmaceutical secondary standard, certified reference material
Synonym(s):
1-(4-Chlorophenyl)-3-cyanoguanidine
Empirical Formula (Hill Notation):
C8H7ClN4
CAS Number:
Molecular Weight:
194.62

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade