Skip to Content
Merck
Pravastatin 1,1,3,3-tetramethylbutylamine certified reference material, pharmaceutical secondary standard

Pravastatin 1,1,3,3-tetramethylbutylamine

Pravastatin 1,1,3,3-tetramethylbutylamine certified reference material, pharmaceutical secondary standard
Empirical Formula (Hill Notation):
C23H36O7 · C8H19N
CAS Number:
Molecular Weight:
553.77

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade