![Plastic additive 03 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/368/862/aa35923f-69bb-4422-b6c5-544be1b0445b/640/aa35923f-69bb-4422-b6c5-544be1b0445b.png)
Plastic additive 03
![Plastic additive 03 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/368/862/aa35923f-69bb-4422-b6c5-544be1b0445b/640/aa35923f-69bb-4422-b6c5-544be1b0445b.png)
Synonym(s):
N,N′-Ethylenebis(stearamide)
Linear Formula:
[CH3(CH2)16CONHCH2-]2
CAS Number:
Molecular Weight:
593.02
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing