Skip to Content
Merck
Phenethyl alcohol ≥99%, FCC, FG

Phenethyl alcohol

Phenethyl alcohol ≥99%, FCC, FG
Synonym(s):
2-Phenylethanol, β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA
Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing