
PEP TCHA

Synonym(s):
Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt, 2-(Phosphonooxy)-2-propenoic acid tri(cyclohexylammonium) salt, PEP-3CHA, Phosphoenolpyruvic acid tris(cyclohexylamine) salt
Empirical Formula (Hill Notation):
C3H5O6P · 3C6H13N
CAS Number:
Molecular Weight:
465.56