Skip to Content
JP
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pellicon
®
XL Cassette with Ultracel
®
Membrane
Products
Product Category
filters (5)
Brand
Millipore (5)
pH Value
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Membrane Material
MWCO (kDa)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
PXC030C50
Pellicon
®
XL Cassette with Ultracel
®
Membrane,
filtration area 50 cm
2
, NMWCO 30 kDa
Expand
View Pricing
PXC300C50
Pellicon
®
XL Cassette with Ultracel
®
Membrane,
filtration area 50 cm
2
, NMWCO 300 kDa
Expand
View Pricing
PXC010C50
Pellicon
®
XL Cassette with Ultracel
®
Membrane,
filtration area 50 cm
2
, NMWCO 10 kDa
Expand
View Pricing
PXC005C50
Pellicon
®
XL Cassette with Ultracel
®
Membrane,
filtration area 50 cm
2
, NMWCO 5 kDa
Expand
View Pricing
PXC01MC50
Pellicon
®
XL Cassette with Ultracel
®
Membrane,
filtration area 50 cm
2
, NMWCO 1000 kDa
Expand
View Pricing