![Parteck® Delta M (Mannitol) EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, E 421](/deepweb/assets/sigmaaldrich/product/images/311/048/1468862b-2c09-408f-810c-26967205badf/640/1468862b-2c09-408f-810c-26967205badf.jpg)
Parteck® Delta M (Mannitol)
![Parteck® Delta M (Mannitol) EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, E 421](/deepweb/assets/sigmaaldrich/product/images/311/048/1468862b-2c09-408f-810c-26967205badf/640/1468862b-2c09-408f-810c-26967205badf.jpg)
Synonym(s):
D-Mannitol
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing