PAMAM dendrimer
Linear Formula:
NH2(CH2)2NH2]:(G=0);dendri PAMAM(NH2)4
CAS Number:
Molecular Weight:
516.68
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing