Skip to Content
Merck
Palmitoleic acid analytical standard

Palmitoleic acid

Palmitoleic acid analytical standard
Synonym(s):
cis-9-Hexadecenoic acid
Linear Formula:
CH3(CH2)5CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
254.41
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing