![Oleyl Oleate 95 United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/507/98416a58-8ddd-41a1-8e43-4cbc81974ba1/640/98416a58-8ddd-41a1-8e43-4cbc81974ba1.png)
Oleyl Oleate 95
![Oleyl Oleate 95 United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/507/98416a58-8ddd-41a1-8e43-4cbc81974ba1/640/98416a58-8ddd-41a1-8e43-4cbc81974ba1.png)
Synonym(s):
Oleyl oleate, Oleic acid oleyl ester
Empirical Formula (Hill Notation):
C36H68O2
CAS Number:
Molecular Weight:
532.92
Products
Formula Weight
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing