Skip to Content
Merck
Octanoic acid-1-13C Extra 99 atom % 13C

Octanoic acid-1-13C Extra

Octanoic acid-1-13C Extra 99 atom % 13C
Synonym(s):
Octanoic acid-1-13C, Caprylic acid-1-13C
Linear Formula:
CH3(CH2)613CO2H
CAS Number:
Molecular Weight:
145.20

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing