Skip to Content
Merck
Obacunone phyproof® Reference Substance

Obacunone

Obacunone phyproof® Reference Substance
Synonym(s):
Obacunoic acid 3,4-lactone, Tricoccin S3
Empirical Formula (Hill Notation):
C26H30O7
CAS Number:
Molecular Weight:
454.51

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing