Skip to Content
Merck
Nonanal 95%

Nonanal

Nonanal 95%
Synonym(s):
Aldehyde C9, Nonyl aldehyde, Pelargonaldehyde, Nonanal
Linear Formula:
CH3(CH2)7CHO
CAS Number:
Molecular Weight:
142.24

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing