Skip to Content
Merck
Nivalenol in acetonitrile IRMM®, certified reference material

Nivalenol in acetonitrile

Nivalenol in acetonitrile IRMM®, certified reference material
Synonym(s):
Nivalenol solution, 3α,4β,7α, 15-Tetrahydroxy-12,13-epoxytrichothec-9-en-8-one, NIV
Empirical Formula (Hill Notation):
C15H20O7
CAS Number:
Molecular Weight:
312.32
EC Number:

Products

Product Category
Formula Weight
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing