
Nicotinamide Impurity F

Synonym(s):
1-Methylnicotinamide chloride, 3-Carbamyl-1-methylpyridinium chloride, MNA, N-Methylnicotinic acid amide chloride, N1-Methylnicotinamide chloride
Empirical Formula (Hill Notation):
C7H9ClN2O
CAS Number:
Molecular Weight:
172.61
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing