![Nevirapine Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/263/203/39b88521-a390-45cf-9fb8-eed9bbfeff46/640/39b88521-a390-45cf-9fb8-eed9bbfeff46.png)
Nevirapine
![Nevirapine Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/263/203/39b88521-a390-45cf-9fb8-eed9bbfeff46/640/39b88521-a390-45cf-9fb8-eed9bbfeff46.png)
Synonym(s):
11-Cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one, Nevirapine
Empirical Formula (Hill Notation):
C15H14N4O
CAS Number:
Molecular Weight:
266.30
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3