Skip to Content
Merck
vibrant-m

Netilmicin sulfate for LC assay

vibrant-m
Synonym(s):
Netilmicin sulfate salt, 1-N-Ethylsisomicin
Linear Formula:
C21H41N5O7 · 5/2H2SO4
CAS Number:
Molecular Weight:
720.78
EC Number:

Products

Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1