Skip to Content
Merck
N-Acetyl-D,L-Methionine Pharmaceutical Secondary Standard; Certified Reference Material

N-Acetyl-D,L-Methionine

N-Acetyl-D,L-Methionine Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
N-Acetyl-DL-methionine, Methionamine
Linear Formula:
CH3SCH2CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
191.25

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade