Montelukast for peak identification
Synonym(s):
Montelukast sodium
Empirical Formula (Hill Notation):
C35H35ClNO3S·Na
CAS Number:
Molecular Weight:
608.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing