Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM) antibody produced in mouse
Synonym(s):
Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM)
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Research Area
Conjugate
Post Translational Modification
Non-reactive Species
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing