![Mexiletine impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/288/561/66911316-eb4d-4a3d-949d-b20e92e9c3c0/640/66911316-eb4d-4a3d-949d-b20e92e9c3c0.png)
Mexiletine impurity A
![Mexiletine impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/288/561/66911316-eb4d-4a3d-949d-b20e92e9c3c0/640/66911316-eb4d-4a3d-949d-b20e92e9c3c0.png)
Synonym(s):
2,6-Dimethylphenol, 2-Hydroxy-m-xylene, vic.-m-Xylenol
Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing