Methyl red (C.I. 13020)
Synonym(s):
Methyl red (C.I. 13020)
Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
Products
Brand
Color
Formula Weight
Physical Form
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing