![Methylprednisolone Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/340/007/cb2eef1d-cc49-4327-b563-0ab2d6864e8b/640/cb2eef1d-cc49-4327-b563-0ab2d6864e8b.jpg)
Methylprednisolone Related Compound C
![Methylprednisolone Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/340/007/cb2eef1d-cc49-4327-b563-0ab2d6864e8b/640/cb2eef1d-cc49-4327-b563-0ab2d6864e8b.jpg)
Synonym(s):
11β-Hydroxy-6α-methylandrosta-1,4-diene-3,17-dione
Empirical Formula (Hill Notation):
C20H26O3
CAS Number:
Molecular Weight:
314.42
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2