Skip to Content
Merck
Methamidophos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Methamidophos

Methamidophos certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
O,S-Dimethyl phosphoramidothioate
Empirical Formula (Hill Notation):
C2H8NO2PS
CAS Number:
Molecular Weight:
141.13

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique