Skip to Content
Merck
Mecoprop-P PESTANAL®, analytical standard

Mecoprop-P

Mecoprop-P PESTANAL®, analytical standard
Synonym(s):
(R)-2-(4-Chloro-2-methylphenoxy)propionic acid
Empirical Formula (Hill Notation):
C10H11ClO3
CAS Number:
Molecular Weight:
214.65
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing