Skip to Content
Merck
L-Lysine hydrochloride natural, FG

L-Lysine hydrochloride

L-Lysine hydrochloride natural, FG
Synonym(s):
L-Lysine monohydrochloride, (S)-2,6-Diaminohexanoic acid monohydrochloride
Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2