
L-Leucine-1-13C

Linear Formula:
(CH3)2CHCH2CH(NH2)13CO2H
CAS Number:
Molecular Weight:
132.17
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing