![L-Lactate Dehydrogenase (LDHA) from human, recombinant, expressed in E. coli, aqueous solution](/deepweb/assets/sigmaaldrich/product/images/295/218/48f7818e-ed15-41f7-91be-51e9f6cc3df9/640/48f7818e-ed15-41f7-91be-51e9f6cc3df9.jpg)
L-Lactate Dehydrogenase (LDHA)
![L-Lactate Dehydrogenase (LDHA) from human, recombinant, expressed in E. coli, aqueous solution](/deepweb/assets/sigmaaldrich/product/images/295/218/48f7818e-ed15-41f7-91be-51e9f6cc3df9/640/48f7818e-ed15-41f7-91be-51e9f6cc3df9.jpg)
Synonym(s):
Lactic Dehydrogenase, recombinant from E. coli, (S)-Lactate: NAD+ oxidoreductase, L-Lactate Dehydrogenase, Lactate
CAS Number:
Products
Brand
Biological Source
Color
Gene Alias
Physical Form
Recombinant Host
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing