Skip to Content
Merck
Juvenile hormone III ≥65%, liquid, non-sterile

Juvenile hormone III

Juvenile hormone III ≥65%, liquid, non-sterile
Synonym(s):
C16-Juvenile Hormone, JH-III, Manduca hormone, trans,trans-10,11-Epoxy-3,7,11-trimethyl-2,6-dodecadienic acid methyl ester
Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing