
Immobilon® MCE Membrane Filter

Products
Product Category
Brand
Color
Manufacturer
Diameter (mm)
Detection Method
Quality Segment
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing