Skip to Content
Merck
Guanine British Pharmacopoeia (BP) Reference Standard

Guanine

Guanine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-Amino-1,7-dihydro-6H-purin-6-one, 2-Amino-6-hydroxypurine, 2-Aminohypoxanthine
Empirical Formula (Hill Notation):
C5H5N5O
CAS Number:
Molecular Weight:
151.13
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique