Skip to Content
Merck
Glyceryl triheptadecanoate ≥99%

Glyceryl triheptadecanoate

Glyceryl triheptadecanoate ≥99%
Synonym(s):
1,2,3-Triheptadecanoylglycerol, Triheptadecanoin
Linear Formula:
[CH3(CH2)15COOCH2]2CHOCO(CH2)15CH3
CAS Number:
Molecular Weight:
849.40

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing