![Fmoc-Gln(Trt)-OH ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/107/424/36439a36-0c04-453e-a42f-3dc425dabf91/640/36439a36-0c04-453e-a42f-3dc425dabf91.png)
Fmoc-Gln(Trt)-OH
![Fmoc-Gln(Trt)-OH ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/107/424/36439a36-0c04-453e-a42f-3dc425dabf91/640/36439a36-0c04-453e-a42f-3dc425dabf91.png)
Synonym(s):
Nα-Fmoc-Nδ-trityl-L-glutamine, Fmoc-Gln(Trt)-OH
Empirical Formula (Hill Notation):
C39H34N2O5
CAS Number:
Molecular Weight:
610.70
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing