<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
JP
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Custom Products
Digital Platforms & Software
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Ethylene glycol monophenyl ether
Synonym(s):
2-Phenoxyethanol, Phenylglycol, Ethylene glycol monophenyl ether
Linear Formula:
C
6
H
5
OCH
2
CH
2
OH
CAS Number:
122-99-6
Molecular Weight:
138.16
EC Number:
204-589-7
Products
Shipping
Ships Today (1)
Product Category
alcohol monomers (1)
monomers (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
484318
Ethylene glycol monophenyl ether,
≥90%
Expand
View Pricing
8.07291
Ethylene glycol monophenyl ether,
for synthesis
Expand
View Pricing
09-1755
Ethylene glycol monophenyl ether,
SAJ first grade, ≥95.0%
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
日本語 - JA
English - EN
Learn More